Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-32
Entrez Gene ID
188687
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_T21C9.7
Chromosome Location
Chr.V: 10587066 - 10588794 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073315.3 NP_505716.2 2 - 3 376 121

View Full Product Details