Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
deg-3
Entrez Gene ID
3565200
Gene Name
Acetylcholine receptor subunit alpha-type deg-3
Gene Aliases
CELE_K03B8.9
Chromosome Location
Chr.V: 11412865 - 11415159 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073496.6 NP_505897.1 2 - 3 290 132
GenBank mRNA U19747.1 - 2 - 3 312 132

View Full Product Details