The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_073525.8 | NP_505926.2 | 8 - 9 | 1272 | 135 |
NM_073526.5 | NP_505927.2 | 6 - 7 | 1317 | 135 |