Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ape-1
Entrez Gene ID
179601
Gene Name
protein-coding
Gene Aliases
CELE_F46F3.4
Chromosome Location
Chr.V: 11675938 - 11685274 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269424.2 NP_001256353.1 1 - 2 85 70

View Full Product Details