Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sulp-4
Entrez Gene ID
179626
Gene Name
STAS domain-containing protein
Gene Aliases
CELE_K12G11.1
Chromosome Location
Chr.V: 11875360 - 11879141 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073588.5 NP_505989.1 4 - 5 667 85
GenBank mRNA AY887910.1 - 4 - 5 689 85

View Full Product Details