Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mif-2
Entrez Gene ID
179638
Gene Name
protein-coding
Gene Aliases
CELE_C52E4.2
Chromosome Location
Chr.V: 11976777 - 11978459 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380778.1 3 - 4 185 76
NM_001380779.1 NP_001366841.1 3 - 4 240 76

View Full Product Details