Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-46
Entrez Gene ID
179782
Gene Name
Molybdopterin molybdenumtransferase
Gene Aliases
CELE_R186.4
Chromosome Location
Chr.V: 12968855 - 12970730 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073845.5 NP_506246.1 3 - 4 452 151
GenBank mRNA AF042069.1 - 3 - 4 452 151

View Full Product Details