Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
try-10
Entrez Gene ID
179787
Gene Name
protein-coding
Gene Aliases
CELE_F15B9.5
Chromosome Location
Chr.V: 13010275 - 13013286 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269546.3 NP_001256475.1 7 - 8 835 96
NM_001269547.3 NP_001256476.1 1 - 2 87 96
NM_001368472.3 NP_001355438.1 6 - 6 633 96

View Full Product Details