Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-9
Entrez Gene ID
186172
Gene Name
Serpentine Receptor, class V
Gene Aliases
CELE_F53F1.8
Chromosome Location
Chr.V: 13421140 - 13423147 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356747.2 NP_001343570.1 4 - 5 633 73

View Full Product Details