Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
avr-15
Entrez Gene ID
179834
Gene Name
protein-coding
Gene Aliases
CELE_R11G10.1
Chromosome Location
Chr.V: 13498780 - 13511718 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028905.8 NP_001024076.1 2 - 3 287 84

View Full Product Details