Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-32
Entrez Gene ID
192075
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_F53C11.6
Chromosome Location
Chr.V: 13796641 - 13800577 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074015.3 NP_506416.2 6 - 7 866 107

View Full Product Details