Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sptl-3
Entrez Gene ID
179884
Gene Name
protein-coding
Gene Aliases
CELE_T22G5.5
Chromosome Location
Chr.V: 13893325 - 13896417 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269618.3 NP_001256547.1 6 - 7 882 106
NM_001269619.2 NP_001256548.1 5 - 6 865 106

View Full Product Details