Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gnrr-2
Entrez Gene ID
182647
Gene Name
protein-coding
Gene Aliases
CELE_C15H11.2
Chromosome Location
Chr.V: 14409074 - 14412003 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269654.4 NP_001256583.1 4 - 5 701 81
NM_001269655.4 NP_001256584.2 2 - 3 331 81

View Full Product Details