Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-55
Entrez Gene ID
184218
Gene Name
C-type LECtin
Gene Aliases
CELE_F08H9.9
Chromosome Location
Chr.V: 14479279 - 14482317 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074190.3 NP_506591.1 1 - 2 58 75

View Full Product Details