Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pes-9
Entrez Gene ID
179963
Gene Name
M20_dimer domain-containing protein
Gene Aliases
CELE_R11H6.1
Chromosome Location
Chr.V: 14595313 - 14597034 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074209.8 NP_506610.1 1 - 2 304 88

View Full Product Details