Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
haf-7
Entrez Gene ID
190115
Gene Name
HAlF transporter (PGP related)
Gene Aliases
CELE_Y50E8A.16
Chromosome Location
Chr.V: 14773258 - 14777697 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074244.4 NP_506645.1 1 - 2 645 64

View Full Product Details