Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-3
Entrez Gene ID
182084
Gene Name
J domain-containing protein
Gene Aliases
CELE_C01G10.12
Chromosome Location
Chr.V: 15068678 - 15069507 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074310.4 NP_506711.1 1 - 2 144 99

View Full Product Details