Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-96
Entrez Gene ID
188391
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T10H4.2
Chromosome Location
Chr.V: 15292490 - 15293965 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074388.1 NP_506789.1 2 - 3 284 124

View Full Product Details