Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyb-3
Entrez Gene ID
180040
Gene Name
protein-coding
Gene Aliases
CELE_T06E6.2
Chromosome Location
Chr.V: 15393906 - 15397351 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074424.9 NP_506825.4 2 - 3 1575 67
GenBank mRNA U21282.1 - 1 - 2 64 67

View Full Product Details