Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-9
Entrez Gene ID
186433
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F57A10.1
Chromosome Location
Chr.V: 15758749 - 15760465 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074525.3 NP_506926.1 2 - 3 792 105

View Full Product Details