Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-69
Entrez Gene ID
180060
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_F54B8.3
Chromosome Location
Chr.V: 15814232 - 15815971 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074547.6 NP_506948.1 3 - 4 1292 79

View Full Product Details