Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-57
Entrez Gene ID
188163
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T06C12.2
Chromosome Location
Chr.V: 15870656 - 15872398 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074564.2 NP_506965.1 1 - 2 1007 122

View Full Product Details