Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-56
Entrez Gene ID
188164
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T06C12.3
Chromosome Location
Chr.V: 15872573 - 15873770 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074565.1 NP_506966.1 2 - 3 928 132

View Full Product Details