Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-57
Entrez Gene ID
185365
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F36G9.1
Chromosome Location
Chr.V: 15953595 - 15955032 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074596.2 NP_506997.2 2 - 3 468 110

View Full Product Details