Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-34
Entrez Gene ID
184400
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F13A7.3
Chromosome Location
Chr.V: 16372417 - 16374776 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074734.2 NP_507135.2 4 - 5 757 94

View Full Product Details