Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-14
Entrez Gene ID
188293
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T08G3.2
Chromosome Location
Chr.V: 16453393 - 16455165 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074760.1 NP_507161.1 4 - 5 941 124

View Full Product Details