Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-70
Entrez Gene ID
180217
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_Y51A2D.17
Chromosome Location
Chr.V: 18616587 - 18620243 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075235.6 NP_507636.1 4 - 5 514 58

View Full Product Details