Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srw-39
Entrez Gene ID
184662
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F19B2.3
Chromosome Location
Chr.V: 20169480 - 20172041 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356791.1 NP_001343561.1 2 - 3 455 110

View Full Product Details