Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srd-70
Entrez Gene ID
185988
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_F48F5.4
Chromosome Location
Chr.V: 20454111 - 20456487 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269992.2 NP_001256921.1 2 - 3 388 66
NR_070284.1 2 - 3 288 66

View Full Product Details