Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-25
Entrez Gene ID
180356
Gene Name
protein-coding
Gene Aliases
CELE_W07A8.3
Chromosome Location
Chr.V: 20729948 - 20744346 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270018.2 NP_001256947.1 3 - 4 359 103
NM_001373209.3 NP_001360578.1 4 - 4 366 103
GenBank mRNA AF293972.1 - 3 - 4 379 103

View Full Product Details