Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trap-2
Entrez Gene ID
180422
Gene Name
Translocon-associated protein subunit beta
Gene Aliases
CELE_T04G9.5
Chromosome Location
Chr.X: 767023 - 768069 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075749.8 NP_508150.1 1 - 2 73 67

View Full Product Details