Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ile-2
Entrez Gene ID
180423
Gene Name
L-type lectin-like domain-containing protein
Gene Aliases
CELE_T04G9.3
Chromosome Location
Chr.X: 768596 - 770776 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075750.8 NP_508151.1 5 - 6 840 91

View Full Product Details