Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pdf-2
Entrez Gene ID
185966
Gene Name
PDF (arthropod Pigment Dispersing Factor) homolog
Gene Aliases
CELE_F48B9.4
Chromosome Location
Chr.X: 2163384 - 2164069 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075996.3 NP_508397.2 1 - 2 101 66
GenBank mRNA EF141321.1 - 1 - 2 101 66

View Full Product Details