Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-31
Entrez Gene ID
191655
Gene Name
protein-coding
Gene Aliases
CELE_T07D1.1
Chromosome Location
Chr.X: 2419955 - 2427504 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029718.2 NP_001024889.1 7 - 8 897 133
NM_001029719.1 NP_001024890.1 5 - 6 525 133
NM_001330893.1 NP_001317860.1 6 - 7 786 133
GenBank mRNA AY275182.1 - 7 - 8 897 133
AY275183.1 - 5 - 6 525 133

View Full Product Details