Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mai-1
- Entrez Gene ID
-
3565232
- Gene Name
- ATPase inhibitor mai-1, mitochondrial
- Gene Aliases
-
CELE_K10B3.9
- Chromosome Location
-
Chr.X: 3113569 - 3116589 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076135.7 |
NP_508536.1 |
2 - 3 |
657 |
66 |
GenBank mRNA |
L12348.1 |
- |
2 - 3 |
661 |
66 |
- Gene Symbol
- gpd-2
- Entrez Gene ID
-
3565504
- Gene Name
- Glyceraldehyde-3-phosphate dehydrogenase 2
- Gene Aliases
-
CELE_K10B3.8
- Chromosome Location
-
Chr.X: 3112291 - 3116581 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383540.1 |
|
2 - 3 |
653 |
66 |
View Full Product Details