Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fax-1
Entrez Gene ID
180609
Gene Name
protein-coding
Gene Aliases
CELE_F56E3.4
Chromosome Location
Chr.X: 3196064 - 3199349 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277024.2 NP_001263953.1 2 - 3 187 63
NM_076146.4 NP_508547.1 2 - 3 159 63
GenBank mRNA AF176087.1 - 2 - 3 159 63

View Full Product Details