Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-78
Entrez Gene ID
180631
Gene Name
Actin-interacting protein 1
Gene Aliases
CELE_C04F6.4
Chromosome Location
Chr.X: 3408664 - 3411615 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029185.3 NP_001024356.1 5 - 6 1731 141
NR_136349.1 4 - 5 1778 141
GenBank mRNA AF324437.1 - 5 - 6 1740 141

View Full Product Details