Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cgr-1
Entrez Gene ID
180650
Gene Name
CRAL/TRIO and GOLD domain suppressor of activated Ras
Gene Aliases
CELE_T27A10.7
Chromosome Location
Chr.X: 3597271 - 3601959 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076217.6 NP_508618.2 4 - 5 688 68

View Full Product Details