Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-16
Entrez Gene ID
180756
Gene Name
protein-coding
Gene Aliases
CELE_R08E3.4
Chromosome Location
Chr.X: 4805998 - 4811542 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029679.5 NP_001024850.1 7 - 8 755 96
NM_001029680.5 NP_001024851.1 7 - 8 761 96

View Full Product Details