Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpyd-1
Entrez Gene ID
180818
Gene Name
Dihydropyrimidine dehydrogenase [NADP(+)]
Gene Aliases
CELE_C25F6.3
Chromosome Location
Chr.X: 5467870 - 5472684 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076526.6 NP_508927.2 12 - 13 3021 83

View Full Product Details