Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-97
Entrez Gene ID
180827
Gene Name
LIM domain-containing protein unc-97
Gene Aliases
CELE_F14D12.2
Chromosome Location
Chr.X: 5593133 - 5594965 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076542.8 NP_508943.3 6 - 7 919 91
GenBank mRNA AF035583.1 - 6 - 7 921 91

View Full Product Details