Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
klp-13
Entrez Gene ID
180865
Gene Name
Kinesin-like protein
Gene Aliases
CELE_F22F4.3
Chromosome Location
Chr.X: 5984989 - 5992151 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373314.3 NP_001360738.1 4 - 4 424 97

View Full Product Details