Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abts-4
Entrez Gene ID
180943
Gene Name
protein-coding
Gene Aliases
CELE_R03E9.3
Chromosome Location
Chr.X: 6747296 - 6761744 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029655.6 NP_001024826.2 12 - 13 1784 106
NM_001129697.5 NP_001123169.1 12 - 13 1784 106
NM_001129698.4 NP_001123170.1 12 - 13 1742 106
NM_001373337.3 NP_001360088.1 13 - 13 1790 106
GenBank mRNA AY887906.1 - 12 - 13 1806 106
AY954522.1 - 12 - 13 1806 106

View Full Product Details