Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gei-15
Entrez Gene ID
24104769
Gene Name
GEX Interacting protein
Gene Aliases
CELE_M03A8.4
Chromosome Location
Chr.X: 6820005 - 6823274 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307909.3 NP_001294838.1 4 - 5 500 79

View Full Product Details