The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001330903.3 | NP_001317892.1 | 16 - 17 | 2986 | 95 |
NM_001373345.2 | NP_001360631.1 | 18 - 19 | 3268 | 95 | |
NM_001373346.2 | NP_001360716.1 | 17 - 17 | 2998 | 95 | |
NM_076800.7 | NP_509201.3 | 15 - 16 | 2768 | 95 |