Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bar-1
Entrez Gene ID
180982
Gene Name
Beta-catenin/armadillo-related protein 1
Gene Aliases
CELE_C54D1.6
Chromosome Location
Chr.X: 7166005 - 7170625 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076805.8 NP_509206.1 8 - 9 1004 69
GenBank mRNA AF063646.1 - 8 - 9 1029 69

View Full Product Details