Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdpn-1
Entrez Gene ID
181058
Gene Name
protein-coding
Gene Aliases
CELE_F45E1.7
Chromosome Location
Chr.X: 7988931 - 7998229 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381034.1 NP_001367622.1 6 - 7 646 66
NM_001381035.1 NP_001367621.1 6 - 7 780 66

View Full Product Details