Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-29
Entrez Gene ID
181081
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_F13B9.6
Chromosome Location
Chr.X: 8262420 - 8271021 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076994.5 NP_509395.2 3 - 4 268 110

View Full Product Details