Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
utx-1
Entrez Gene ID
181110
Gene Name
protein-coding
Gene Aliases
CELE_D2021.1
Chromosome Location
Chr.X: 8557146 - 8563238 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322626.3 NP_001309685.1 6 - 7 1035 92

View Full Product Details