Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-12
Entrez Gene ID
181157
Gene Name
Guanine nucleotide-binding protein alpha-12 subunit
Gene Aliases
CELE_F18G5.3
Chromosome Location
Chr.X: 9243557 - 9249107 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077156.6 NP_509557.1 6 - 7 657 136
GenBank mRNA AY008134.1 - 5 - 6 585 136

View Full Product Details